siRNA Details
Gene Symbol: Naip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010871.2 | NP_035001.2 | 9 | 2050 | ||
GenBank mRNA | AF135494.1 | - | 2174 | |||
AF381772.1 | - | 9 | 2050 | |||
BC141346.1 | - | 10 | 2116 |
Gene Symbol: Naip7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021545.1 | NP_067520.1 | 2184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
