siRNA Details
Gene Symbol: Ptprz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081306.1 | NP_001074775.1 | 2 | 422 | ||
GenBank mRNA | AJ133130.1 | - | 2 | 202 | ||
AJ428208.1 | - | 97 | ||||
AK081698.1 | - | 2 | 423 | |||
BC151071.1 | - | 2 | 147 | 9087984 | ||
BC157965.1 | - | 2 | 147 | 9088134 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
