siRNA Details
Gene Symbol: Ccl27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048179.1 | NP_001041644.1 | 5 | 342 | ||
NM_011336.1 | NP_035466.1 | 2 | 176 | |||
GenBank mRNA | AB013398.1 | - | 2 | 176 | ||
AF082392.1 | - | 2 | 154 | |||
AF099931.1 | - | 2 | 164 | |||
AK005398.1 | - | 3 | 316 | |||
AK146066.1 | - | 2 | 1680 | |||
AY744154.1 | - | 1 | 295 | |||
AY744155.1 | - | 1 | 295 | |||
BC107226.2 | - | 2 | 176 | 40051915 | ||
CA451425.1 | - | 5 | 342 |
Gene Symbol: Ccl27b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199959.1 | NP_001186888.1 | 2 | 188 | ||
NM_001199960.1 | NP_001186889.1 | 2 | 475 | |||
NM_001199961.1 | NP_001186890.1 | 404 | ||||
GenBank mRNA | BY705727.1 | - | 2 | 188 |
Gene Symbol: Gm13306 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164046.1 | NP_001157518.1 | 2 | 188 | ||
NM_001199962.1 | NP_001186891.1 | 5 | 404 | |||
NM_001199963.1 | NP_001186892.1 | 2 | 475 | |||
GenBank mRNA | AK005520.1 | - | 2 | 188 | ||
BC028511.1 | - | 2 | 156 | 3471454 | ||
BY613083.1 | - | 2 | 144 | |||
BY652838.1 | - | 1 | 136 |
Gene Symbol: Gm2506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199965.1 | NP_001186894.1 | 2 | 188 | ||
NM_001199966.1 | NP_001186895.1 | 2 | 475 | |||
GenBank mRNA | BE632104.1 | - | 2 | 166 |
Gene Symbol: LOC100861978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267702.1 | NP_001254631.1 | 5 | 404 | ||
NM_001267703.1 | NP_001254632.1 | 2 | 475 | |||
NM_001267704.1 | NP_001254633.1 | 2 | 188 |
