siRNA Details
Gene Symbol: AK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199853.1 | NP_001186782.1 | 2 | 116 | ||
NM_001199855.1 | NP_001186784.1 | 2 | 123 | |||
NM_001199856.1 | NP_001186785.1 | 2 | 189 | |||
NM_016282.3 | NP_057366.2 | 2 | 417 | |||
GenBank mRNA | AF086913.1 | - | 2 | 187 | ||
AF100785.1 | - | 2 | 195 | |||
AF183419.1 | - | 2 | 116 | |||
AK001553.1 | - | 2 | 335 | |||
AK001951.1 | - | 2 | 307 | |||
AK027534.1 | - | 2 | 209 | |||
AK098205.1 | - | 2 | 1613 | |||
AK124824.1 | - | 2 | 957 | |||
BC013771.1 | - | 2 | 251 | 4385034 | ||
BI550730.1 | - | 2 | 268 | |||
BI601245.1 | - | 2 | 422 | |||
DA570364.1 | - | 2 | 196 | |||
DA718987.1 | - | 2 | 123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
