siRNA Details
Gene Symbol: Hmga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010441.2 | NP_034571.1 | 4, 5 | 1162 | ||
GenBank mRNA | AK028346.1 | - | 4, 5 | 492 | ||
AK142059.1 | - | 4, 5 | 524 | |||
BC052158.1 | - | 4, 5 | 696 | 6506864 | ||
BC085085.1 | - | 4, 5 | 463 | 6334495 | ||
X58380.1 | - | 4, 5 | 376 |

More Information
Other Species Targeted

Gene Ontology Categories:
