siRNA Details
Gene Symbol: Ptprz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081306.1 | NP_001074775.1 | 2, 3 | 457 | ||
GenBank mRNA | AJ133130.1 | - | 2, 3 | 237 | ||
AJ428208.1 | - | 132 | ||||
AK081698.1 | - | 2, 3 | 458 | |||
BC151071.1 | - | 2, 3 | 182 | 9087984 | ||
BC157965.1 | - | 2, 3 | 182 | 9088134 |

More Information
Other Species Targeted

Gene Ontology Categories:
