siRNA Details
Gene Symbol: Pla2g5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122954.1 | NP_001116426.1 | 4 | 535 | ||
NM_011110.4 | NP_035240.3 | 3 | 377 | |||
GenBank mRNA | AK157266.1 | - | 3 | 201 | ||
AK161336.1 | - | 3 | 404 | |||
BC030899.1 | - | 3 | 337 | 4502326 | ||
U66873.1 | - | 2 | 109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
