siRNA Details
Gene Symbol: Syncrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284328.1 | NP_001271257.1 | 8 | 1221 | ||
NM_019666.2 | NP_062640.2 | 8 | 1221 | |||
NM_019796.5 | NP_062770.1 | 8 | 1221 | |||
GenBank mRNA | AB035725.1 | - | 8 | 1020 | ||
AF408434.1 | - | 6 | 674 | |||
AK034845.1 | - | 8 | 1033 | |||
AK077588.1 | - | 8 | 970 | |||
AK078158.1 | - | 6 | 654 | |||
AK083398.1 | - | 7 | 812 | |||
AK162448.1 | - | 2 | 103 | |||
BC041148.1 | - | 8 | 1066 | 4946652 | ||
BC055863.1 | - | 8 | 999 | |||
BC058807.1 | - | 8 | 1088 | |||
BC080309.1 | - | 8 | 924 | 5716184 | ||
BC108363.1 | - | 8 | 955 | 4239809 | ||
CN529994.1 | - | 5 | 555 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001047916, XM_006243519, XM_006243515, XM_006243517, XM_006243516, and others.
Gene Ontology Categories:
