siRNA Details
Gene Symbol: Rrbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024281.2 | NP_077243.2 | 2 | 435 | ||
NM_133626.2 | NP_598329.1 | 2 | 435 | |||
GenBank mRNA | AF273683.1 | - | 358 | |||
AF273684.1 | - | 2 | 358 | |||
AF273685.1 | - | 2 | 358 | |||
AF273686.1 | - | 2 | 358 | |||
AF273687.1 | - | 2 | 358 | |||
AF273688.1 | - | 2 | 358 | |||
AF273689.1 | - | 2 | 358 | |||
AF273690.1 | - | 2 | 358 | |||
AF273691.1 | - | 2 | 358 | |||
AF273692.1 | - | 2 | 358 | |||
AF273693.1 | - | 2 | 358 | |||
AK039114.1 | - | 2 | 435 | |||
BC002105.1 | - | 2 | 345 | |||
BC031452.1 | - | 2 | 424 |

More Information
Other Species Targeted

Gene Ontology Categories:
