siRNA Details
Gene Symbol: Aph1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177583.4 | NP_808251.1 | 2 | 326 | ||
GenBank mRNA | AK028521.1 | - | 2 | 340 | ||
AK049828.1 | - | 2 | 327 | |||
AK154847.1 | - | 2 | 394 | |||
AK162585.1 | - | 2 | 308 | |||
BC079659.1 | - | 2 | 358 | 6853991 |
Gene Symbol: Aph1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026674.3 | NP_080950.1 | 2 | 376 | ||
GenBank mRNA | AB197123.1 | - | 2 | 196 | ||
AK002310.1 | - | 2 | 342 | |||
AK080464.1 | - | 1 | 323 | |||
AK155147.1 | - | 2 | 390 | |||
BC050923.1 | - | 2 | 376 | 6410104 | ||
BC063254.1 | - | 2 | 376 | 6410104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
