siRNA Details
Gene Symbol: Rprd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081293.1 | NP_001074762.1 | 1 | 337 | ||
GenBank mRNA | AK012860.1 | - | 1 | 212 | ||
AK015972.1 | - | 1 | 248 | |||
AK083674.1 | - | 1 | 160 | |||
AK087157.1 | - | 1 | 163 | |||
AK129150.1 | - | 1 | 16 | |||
AK154692.1 | - | 1 | 143 | |||
BC067054.1 | - | 1 | 142 | 30542928 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
