siRNA Details
Gene Symbol: Uxs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026430.3 | NP_080706.1 | 3 | 140 | ||
GenBank mRNA | AF399958.1 | - | 3 | 140 | ||
AK005536.1 | - | 3 | 174 | |||
AK075806.1 | - | 3 | 192 | |||
AK076166.1 | - | 3 | 194 | |||
AK152376.1 | - | 3 | 195 | |||
BC037049.1 | - | 3 | 186 | 4935237 |
