siRNA Details
Gene Symbol: Aph1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026674.3 | NP_080950.1 | 3 | 413 | ||
GenBank mRNA | AB197123.1 | - | 3 | 233 | ||
AK002310.1 | - | 3 | 379 | |||
AK155147.1 | - | 3 | 427 | |||
BC050923.1 | - | 3 | 413 | 6410104 | ||
BC063254.1 | - | 3 | 413 | 6410104 |
Gene Symbol: Aph1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177583.4 | NP_808251.1 | 3 | 363 | ||
GenBank mRNA | AK028521.1 | - | 3 | 377 | ||
AK049828.1 | - | 3 | 364 | |||
AK154847.1 | - | 3 | 431 | |||
AK162585.1 | - | 3 | 345 | |||
BC079659.1 | - | 3 | 395 | 6853991 |

More Information
Other Species Targeted

Gene Ontology Categories:
