siRNA Details
Gene Symbol: Ak6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027592.3 | NP_081868.1 | 3, 4 | 257 | ||
GenBank mRNA | AK014903.1 | - | 2, 3 | 179 | ||
BC019453.1 | - | 3, 4 | 208 | 4024616 | ||
BG084768.2 | - | 1, 2 | 44 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
