siRNA Details
Gene Symbol: A230077H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038939.1 | - | 1757 |
Gene Symbol: 4930555F03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042788.1 | - | 3 | 511 |
Gene Symbol: 4931432E15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016500.1 | - | 1 | 1618 |
Gene Symbol: 6530403H02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032656.1 | - | 14 | 2225 |
Gene Symbol: 9430047G12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034849.1 | - | 1 | 38 |
Gene Symbol: A130077B15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038082.1 | - | 3 | 1691 |
Gene Symbol: Bora | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC052183.1 | - | 10 | 1813 | 6336038 |
Gene Symbol: Ces2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC044835.1 | - | 12 | 2512 |
Gene Symbol: Cpa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133296.1 | - | 2 | 1811 |
Gene Symbol: Cyb5r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135261.1 | - | 13 | 3025 |
Gene Symbol: Eif2ak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011163.4 | NP_035293.1 | 16 | 2861 | ||
GenBank mRNA | AK009862.1 | - | 1 | 158 | ||
AK028602.1 | - | 16 | 2862 |
Gene Symbol: Fgd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK166883.1 | - | 3 | 2904 |
Gene Symbol: Gm34483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK144767.1 | - | 289 |
Gene Symbol: Gm3871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK148631.1 | - | 1 | 1603 |
Gene Symbol: Gm39182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038453.1 | - | 435 |
Gene Symbol: Hacd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK085898.1 | - | 955 |
Gene Symbol: Heatr5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008221.1 | - | 295 |
Gene Symbol: Jakmip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK048388.1 | - | 1 | 1717 |
Gene Symbol: L1Md-Tf30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035064.1 | - | 1 | 336 |
Gene Symbol: Macrod2os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031201.1 | - | 4 | 1384 |
Gene Symbol: Nuf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023284.3 | NP_075773.2 | 14 | 2343 | ||
GenBank mRNA | AK132467.1 | - | 14 | 2344 |
Gene Symbol: Syne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079686.1 | NP_001073154.1 | 55 | 9283 | ||
NM_022027.2 | NP_071310.2 | 17 | 3237 | |||
GenBank mRNA | AK036828.1 | - | 18 | 3293 | ||
BC041779.1 | - | 16 | 2834 | |||
BC054456.1 | - | 17 | 3237 | |||
BC110426.1 | - | 25 | 4486 | |||
BC110428.1 | - | 17 | 3217 | |||
BC111034.1 | - | 6 | 1033 |
Gene Symbol: Tmem241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028973.1 | - | 663 |
Gene Symbol: Vmn1r58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030739.2 | NP_109664.2 | 2 | 1845 | ||
GenBank mRNA | AF324869.1 | - | 2 | 1844 |
Gene Symbol: Zfp65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145622.2 | NP_663597.1 | 4 | 2335 | ||
GenBank mRNA | AK135182.1 | - | 1 | 840 | ||
BC016248.1 | - | 3 | 2244 | 4527362 |
Gene Symbol: Zrsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038641.1 | - | 2767 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
