siRNA Details
Gene Symbol: Syncrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284328.1 | NP_001271257.1 | 11 | 1983 | ||
NM_019666.2 | NP_062640.2 | 11 | 1983 | |||
NM_019796.5 | NP_062770.1 | 11 | 1983 | |||
GenBank mRNA | AB035725.1 | - | 11 | 1782 | ||
AF408434.1 | - | 9 | 1436 | |||
AK034845.1 | - | 11 | 1795 | |||
AK076026.1 | - | 11 | 1773 | |||
AK077588.1 | - | 11 | 1732 | |||
AK078158.1 | - | 9 | 1416 | |||
AK083398.1 | - | 10 | 1574 | |||
AK162448.1 | - | 5 | 865 | |||
BC004001.1 | - | 1 | 313 | |||
BC041148.1 | - | 11 | 1828 | 4946652 | ||
BC050079.1 | - | 11 | 1756 | 30012104 | ||
BC055863.1 | - | 11 | 1761 | |||
BC058807.1 | - | 11 | 1850 | |||
BC080309.1 | - | 11 | 1686 | 5716184 | ||
BC108363.1 | - | 11 | 1717 | 4239809 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
