siRNA Details
Gene Symbol: Rrbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024281.2 | NP_077243.2 | 2 | 1578 | ||
NM_133626.2 | NP_598329.1 | 2 | 1578 | |||
GenBank mRNA | AF273683.1 | - | 1501 | |||
AF273684.1 | - | 2 | 1501 | |||
AF273685.1 | - | 2 | 1501 | |||
BC031452.1 | - | 2 | 1567 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
