siRNA Details
Gene Symbol: Pink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026880.2 | NP_081156.2 | 2 | 547 | ||
GenBank mRNA | AB053476.1 | - | 2 | 479 | ||
AF316872.1 | - | 2 | 525 | |||
AK050322.1 | - | 2 | 527 | |||
AK155474.1 | - | 2 | 533 | |||
AK165480.1 | - | 2 | 526 | |||
AK187592.1 | - | 2 | 108 | |||
AK188455.1 | - | 1 | 76 | |||
AK215426.1 | - | 2 | 108 | |||
AK220318.1 | - | 2 | 501 | |||
BC018561.1 | - | 2 | 373 | |||
BC018594.1 | - | 2 | 510 | |||
BC067066.1 | - | 2 | 515 | 30542123 | ||
BC069968.1 | - | 3 | 405 |
