siRNA Details
Gene Symbol: Rpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027371.3 | NP_081647.2 | 7, 8 | 907 | ||
GenBank mRNA | AK010079.1 | - | 7, 8 | 904 | ||
AK171079.1 | - | 7, 8 | 896 | |||
BC028802.1 | - | 1, 2 | 142 | |||
BC056165.1 | - | 7, 8 | 858 | 5694049 | ||
BC089369.1 | - | 7, 8 | 892 | 30550759 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
