siRNA Details
Gene Symbol: Sppl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029012.2 | NP_083288.2 | 9 | 1010 | ||
GenBank mRNA | AK014709.1 | - | 9 | 1348 | ||
BC023131.1 | - | 5 | 625 | |||
BC078437.1 | - | 1 | 462 | |||
BC086646.1 | - | 9 | 1375 | 6817386 | ||
CD806412.1 | - | 5 | 627 |
