siRNA Details
Gene Symbol: Sbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081030.2 | NP_001074499.2 | 35 | 4973 | ||
NM_001170561.1 | NP_001164032.1 | 36 | 5051 | |||
GenBank mRNA | AK020972.1 | - | 6 | 614 | ||
AK076039.1 | - | 3 | 218 | |||
AK129485.1 | - | 35 | 4917 | |||
AK147332.1 | - | 35 | 4930 | |||
BC029156.1 | - | 6 | 609 | |||
BC098209.1 | - | 16 | 2316 | |||
BC157935.1 | - | 35 | 4863 | 9088115 | ||
BC158013.1 | - | 36 | 4941 | 9007320 | ||
BC172094.1 | - | 35 | 4863 | 9054380 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
