siRNA Details
Gene Symbol: B4GALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001497.3 | NP_001488.2 | 2, 3 | 845 | ||
XM_005251440.4 | XP_005251497.1 | 2, 3 | 878 | |||
GenBank mRNA | A23697.1 | - | 2, 3 | 662 | ||
AK296928.1 | - | 3, 4 | 544 | |||
AK312797.1 | - | 2, 3 | 684 | |||
AK316294.1 | - | 3, 4 | 517 | |||
BX648438.1 | - | 1, 2 | 160 | |||
D29805.1 | - | 2, 3 | 728 | |||
HG765099.1 | - | 2, 3 | 690 | |||
M13701.1 | - | 1, 2 | 245 | |||
M22921.1 | - | 2, 3 | 728 | |||
U10473.1 | - | 2, 3 | 386 | |||
U10474.1 | - | 2, 3 | 729 | |||
X13223.1 | - | 2, 3 | 728 | |||
X14085.1 | - | 3 | 725 | |||
X55415.1 | - | 2, 3 | 656 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
