siRNA Details
Gene Symbol: VRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308420.1 | NP_001295349.1 | 3, 4 | 331 | ||
NM_016440.3 | NP_057524.3 | 3, 4 | 493 | |||
XM_005258971.3 | XP_005259028.1 | 3, 4 | 820 | |||
XM_006723237.3 | XP_006723300.1 | 3, 4 | 634 | |||
XM_011527023.2 | XP_011525325.1 | 3, 4 | 820 | |||
GenBank mRNA | AB031052.1 | - | 3, 4 | 265 | ||
AF514788.1 | - | 3, 4 | 325 | |||
AK225154.1 | - | 3, 4 | 238 | |||
AK292918.1 | - | 3, 4 | 236 | |||
AK303529.1 | - | 3, 4 | 236 | |||
BC023556.2 | - | 3, 4 | 272 | 4298578 | ||
BC095449.1 | - | 3, 4 | 245 | 30721912 | ||
BG421578.1 | - | 3, 4 | 414 | |||
BP335032.1 | - | 3, 4 | 331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
Gene Ontology Categories:
