siRNA Details
Gene Symbol: Mylk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139300.3 | NP_647461.3 | 31 | 5549 | ||
GenBank mRNA | AF314149.1 | - | 2 | 374 | ||
AK078556.1 | - | 12 | 2226 | |||
AK162008.1 | - | 13 | 1912 | |||
BC034209.1 | - | 6 | 696 | |||
BC037721.1 | - | 16 | 2112 | |||
BC038902.1 | - | 2 | 322 | |||
BC045197.1 | - | 16 | 3032 | |||
BC058610.1 | - | 31 | 5561 | 30056142 | ||
BC145279.1 | - | 31 | 5536 | 9053833 | ||
BC150749.1 | - | 31 | 5536 | 9087660 | ||
BC171954.1 | - | 31 | 5537 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001105874, XM_006248433, XM_008768712, XM_008768711, XM_006248434.
Gene Ontology Categories:
