siRNA Details
Gene Symbol: Rp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146246.3 | NP_666358.2 | 2 | 635 | ||
GenBank mRNA | AJ491325.1 | - | 1 | 437 | ||
AY168340.1 | - | 1 | 416 | |||
BC129887.1 | - | 2 | 560 | 40111348 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
