siRNA Details
Gene Symbol: Adam6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009545.1 | NP_001009545.1 | 1 | 155 | ||
GenBank mRNA | AK029742.1 | - | 1 | 210 | ||
AK076580.1 | - | 1 | 210 | |||
BC111877.1 | - | 1 | 175 | 40059665 |
Gene Symbol: Adam6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174885.3 | NP_777479.2 | 1 | 256 | ||
GenBank mRNA | AY158689.1 | - | 1 | 256 | ||
BC108387.1 | - | 1 | 209 | |||
BC139059.1 | - | 1 | 241 | 8862081 |

More Information
Other Species Targeted

Gene Ontology Categories:
