siRNA Details
Gene Symbol: Tagap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147155.2 | NP_671511.2 | 4 | 598 | ||
GenBank mRNA | AF314818.1 | - | 4 | 598 | ||
BC119550.1 | - | 4 | 598 | |||
BC119551.1 | - | 4 | 598 | |||
BC139797.1 | - | 4 | 596 | |||
BQ555561.1 | - | 3 | 473 |
Gene Symbol: Tagap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145968.2 | NP_666080.1 | 10 | 1361 | ||
GenBank mRNA | BC030886.1 | - | 10 | 1341 | 4483459 | |
BC039780.1 | - | 10 | 1352 | 4483459 | ||
BC150728.1 | - | 10 | 1306 | 9087639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
