siRNA Details
Gene Symbol: Bcor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029510.3 | NP_083786.2 | 5 | 3813 | ||
NM_175045.3 | NP_778210.2 | 5 | 3813 | |||
GenBank mRNA | AY161170.1 | - | 5 | 3128 | ||
AY161172.1 | - | 5 | 3128 | |||
BC058656.1 | - | 5 | 3314 | 6412868 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
