siRNA Details
Gene Symbol: Ifnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177396.1 | NP_796370.1 | 1 | 107 | ||
GenBank mRNA | AY184375.1 | - | 1 | 107 | ||
AY869696.1 | - | 1 | 107 |
Gene Symbol: Ifnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024673.2 | NP_001019844.2 | 1 | 107 | ||
GenBank mRNA | AY869695.1 | - | 1 | 107 | ||
BC150796.1 | - | 1 | 202 | 9087707 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
