siRNA Details
Gene Symbol: Setd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040398.2 | NP_001035488.2 | 8 | 3146 | ||
GenBank mRNA | BC038367.1 | - | 2 | 224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
