siRNA Details
Gene Symbol: Ythdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177680.3 | NP_808348.2 | 10, 11 | 1519 | ||
GenBank mRNA | AB221619.1 | - | 2, 3 | 143 | ||
AI121974.1 | - | 3, 4 | 254 | |||
AK040341.1 | - | 1, 2 | 3636 | |||
AK173312.1 | - | 7, 8 | 782 | |||
BC022697.1 | - | 3, 4 | 312 | |||
BC037593.1 | - | 4, 5 | 412 | |||
BC044650.1 | - | 8, 9 | 1337 | |||
BC048817.1 | - | 3, 4 | 297 | |||
CO424360.1 | - | 5, 6 | 464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
