siRNA Details
Gene Symbol: Kir3dl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177749.3 | NP_808417.2 | 3 | 200 | ||
GenBank mRNA | AY130461.1 | - | 3 | 186 | ||
AY530782.1 | - | 3 | 173 | |||
AY530783.1 | - | 3 | 173 | |||
BC141012.1 | - | 3 | 186 | 9055624 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
