siRNA Details
Gene Symbol: Dock5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177780.3 | NP_808448.2 | 49 | 5444 | ||
GenBank mRNA | BC016533.1 | - | 3 | 646 | ||
BC050992.1 | - | 25 | 2642 | |||
BC158046.1 | - | 49 | 5304 | 9007351 | ||
BC158135.1 | - | 49 | 5304 | 9088200 | ||
BC158138.1 | - | 49 | 5304 | 9088202 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
