siRNA Details
Gene Symbol: Ttll5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081423.2 | NP_001074892.2 | 3 | 384 | ||
GenBank mRNA | AB093278.1 | - | 3 | 317 | ||
AK029125.1 | - | 2 | 2029 | |||
AK046017.1 | - | 3 | 313 | |||
AK085566.1 | - | 3 | 628 | |||
AM690748.1 | - | 2 | 149 | |||
BC035276.1 | - | 2 | 259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
