siRNA Details
Gene Symbol: Agtr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177322.3 | NP_796296.1 | 3 | 1050 | ||
GenBank mRNA | BC036175.1 | - | 3 | 949 | 4989471 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
