siRNA Details
Gene Symbol: Sbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177324.2 | NP_796298.2 | 37 | 5252 | ||
GenBank mRNA | AK029035.1 | - | 4 | 595 | ||
AK029568.1 | - | 8 | 1255 | |||
BC015069.1 | - | 13 | 1968 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
