siRNA Details
Gene Symbol: Parp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177460.4 | NP_803411.3 | 3 | 499 | ||
GenBank mRNA | AK033791.1 | - | 449 | |||
AK133475.1 | - | 3 | 500 | |||
AK166002.1 | - | 2 | 263 | |||
AK169866.1 | - | 3 | 414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
