siRNA Details
Gene Symbol: Ythdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB221619.1 | - | 5, 6 | 419 | ||
AK173312.1 | - | 9, 10 | 3490 | |||
BC022697.1 | - | 6, 7 | 588 | |||
BC037593.1 | - | 7, 8 | 2046 | |||
BC048817.1 | - | 6, 7 | 573 |

More Information
Other Species Targeted

