siRNA Details
Gene Symbol: Fbxw28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177419.1 | NP_001170890.1 | 6 | 729 | ||
NM_001177420.1 | NP_001170891.1 | 6 | 729 | |||
GenBank mRNA | AK143322.1 | - | 6 | 725 | ||
AK162190.1 | - | 6 | 727 | |||
BC084592.1 | - | 6 | 720 |
Gene Symbol: Fbxw14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015793.2 | NP_056608.1 | 729 | |||
GenBank mRNA | AK087709.1 | - | 729 | |||
AK135682.1 | - | 725 | ||||
AK135771.1 | - | 723 | ||||
BC138018.1 | - | 723 | 8861037 |
Gene Symbol: Fbxw19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177703.3 | NP_808371.1 | 6 | 699 | ||
GenBank mRNA | AK087808.1 | - | 6 | 700 |
Gene Symbol: Fbxw20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008428.3 | NP_001008428.1 | 6 | 744 | ||
GenBank mRNA | AK135785.1 | - | 6 | 725 | ||
AK136200.1 | - | 6 | 722 | |||
BC085247.1 | - | 6 | 725 | 30477089 |
Gene Symbol: Fbxw22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014395.2 | NP_001014417.1 | 6 | 723 | ||
GenBank mRNA | AK135808.1 | - | 4 | 440 | ||
AK162068.1 | - | 2 | 231 | |||
BC084593.1 | - | 6 | 727 | 30477093 | ||
CJ052642.1 | - | 4 | 440 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
