siRNA Details
Gene Symbol: Vps37b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177876.4 | NP_808544.1 | 2 | 290 | ||
GenBank mRNA | AK082658.1 | - | 2 | 251 | ||
AK149893.1 | - | 2 | 257 | |||
AK166359.1 | - | 2 | 271 | |||
AK172114.1 | - | 2 | 251 | |||
AK172337.1 | - | 2 | 251 | |||
BC026744.1 | - | 2 | 291 | 4163734 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
