siRNA Details
Gene Symbol: Vmn2r7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175674.3 | NP_783605.2 | 1 | 62 | ||
GenBank mRNA | AK030224.1 | - | 1161 |
Gene Symbol: Vmn2r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104592.1 | NP_001098062.1 | 1 | 62 |
Gene Symbol: Vmn2r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104615.1 | NP_001098085.1 | 1 | 62 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
