siRNA Details
Gene Symbol: Scgb2b27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100464.2 | NP_001093934.1 | 3 | 357 | ||
NM_001276475.1 | NP_001263404.1 | 3 | 605 | |||
GenBank mRNA | AY293279.1 | - | 2 | 212 | ||
BC024677.1 | - | 3 | 281 | 4193111 | ||
BC057695.1 | - | 2 | 1350 | |||
BC127149.1 | - | 298 | 40053058 | |||
GU269237.1 | - | 3 | 281 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
