siRNA Details
Gene Symbol: Slx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177472.5 | NP_803423.2 | 15 | 4954 | ||
GenBank mRNA | AK041127.1 | - | 4 | 1205 | ||
AK042952.1 | - | 4 | 720 | |||
AK173263.1 | - | 14 | 4367 | |||
BC042798.1 | - | 4 | 1625 | |||
BC065125.1 | - | 15 | 4959 | 30534878 | ||
BC082337.1 | - | 8 | 3445 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
