siRNA Details
Gene Symbol: Zfp281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160251.1 | NP_001153723.1 | 2 | 2779 | ||
NM_177643.4 | NP_808311.1 | 2 | 2775 | |||
GenBank mRNA | AK034110.1 | - | 1 | 45 | ||
AK142218.1 | - | 2 | 2692 | |||
AK157189.1 | - | 2 | 2718 | |||
BC003243.1 | - | 2 | 2736 | 3498439 | ||
BC062153.1 | - | 2 | 2729 | 6335572 | ||
CT010200.1 | - | 1 | 2610 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001012030, BC083844.
Gene Ontology Categories:
