siRNA Details
Gene Symbol: Ythdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177680.3 | NP_808348.2 | 15 | 1890 | ||
GenBank mRNA | AB221619.1 | - | 7 | 490 | ||
AK080576.1 | - | 2 | 102 | |||
AK173312.1 | - | 11 | 3561 | |||
BC022697.1 | - | 8 | 659 | |||
BC037593.1 | - | 9 | 2117 | |||
BC044650.1 | - | 11 | 5167 | |||
BC048817.1 | - | 8 | 644 | |||
DV042054.1 | - | 3 | 181 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_133423, XM_006250805, XM_008769987, XM_008769988, XM_008769990, and others.
Gene Ontology Categories:
