siRNA Details
Gene Symbol: Kir3dl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177748.2 | NP_808416.1 | 4, 5 | 668 | ||
GenBank mRNA | AF548540.1 | - | 4, 5 | 668 | ||
AF548541.1 | - | 4, 5 | 668 | |||
AK046171.1 | - | 6, 7 | 867 | |||
AY530784.1 | - | 4, 5 | 668 | |||
BC132013.1 | - | 4, 5 | 717 | |||
BC132015.1 | - | 4, 5 | 717 |
Gene Symbol: Kir3dl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY130461.1 | - | 4, 5 | 681 | ||
AY530783.1 | - | 4, 5 | 668 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
