siRNA Details
Gene Symbol: Dock5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177780.3 | NP_808448.2 | 50 | 5515 | ||
GenBank mRNA | BC016533.1 | - | 4 | 717 | ||
BC050992.1 | - | 26 | 2713 | |||
BC158046.1 | - | 50 | 5375 | 9007351 | ||
BC158135.1 | - | 50 | 5375 | 9088200 | ||
BC158138.1 | - | 50 | 5375 | 9088202 |

More Information
Other Species Targeted

Gene Ontology Categories:
