siRNA Details
Gene Symbol: 3830406C13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284383.1 | NP_001271312.1 | 4 | 286 | ||
NM_001284384.1 | NP_001271313.1 | 4 | 362 | |||
NM_001284385.1 | NP_001271314.1 | 4 | 365 | |||
NM_001284386.1 | NP_001271315.1 | 3 | 205 | |||
NM_001284387.1 | NP_001271316.1 | 3 | 205 | |||
NM_146051.3 | NP_666163.1 | 4 | 289 | |||
NM_178141.3 | NP_835277.1 | 4 | 289 | |||
GenBank mRNA | AK076182.1 | - | 4 | 280 | ||
AK089971.1 | - | 4 | 262 | |||
BC027421.1 | - | 3 | 249 | 4194414 | ||
BC057656.1 | - | 4 | 311 | 4020532 | ||
BC094456.1 | - | 4 | 267 | 5137611 | ||
BE624571.1 | - | 2 | 165 | |||
BQ715343.1 | - | 3 | 247 | |||
BY233185.1 | - | 4 | 275 |
