siRNA Details
Gene Symbol: B430306N03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177083.4 | NP_796057.1 | 2 | 370 | ||
GenBank mRNA | AK036709.1 | - | 2 | 371 | ||
AK046672.1 | - | 371 | ||||
AK133242.1 | - | 2 | 1231 | |||
AK155881.1 | - | 2 | 386 | |||
AK157919.1 | - | 2 | 109 | |||
AK170621.1 | - | 2 | 392 | |||
AK171230.1 | - | 2 | 369 | |||
AK171603.1 | - | 2 | 369 | |||
AK171605.1 | - | 2 | 386 | |||
AY522648.1 | - | 2 | 371 | |||
DQ087179.1 | - | 2 | 123 | |||
DQ087180.1 | - | 2 | 123 |
Gene Symbol: A530064D06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113556.1 | NP_001107028.1 | 2 | 373 | ||
NM_178796.6 | NP_848911.2 | 2 | 373 | |||
GenBank mRNA | AB372005.1 | - | 2 | 150 | ||
AK080114.1 | - | 2 | 334 | |||
AK089248.1 | - | 2 | 387 | |||
AK089332.1 | - | 2 | 334 | |||
AK170607.1 | - | 2 | 392 | |||
AK171433.1 | - | 334 | ||||
BC139185.1 | - | 2 | 208 | 8862207 | ||
BC139186.1 | - | 2 | 208 | 8862208 | ||
BC145572.1 | - | 2 | 208 | 9054138 | ||
EU003111.1 | - | 2 | 104 | |||
EU003112.1 | - | 2 | 104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
