siRNA Details
Gene Symbol: Scaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178923.3 | NP_849254.2 | 4 | 578 | ||
GenBank mRNA | AK045266.1 | - | 4 | 575 | ||
AK048239.1 | - | 4 | 640 | |||
AK173120.1 | - | 4 | 656 | |||
AK219509.1 | - | 2 | 60 | |||
BC053096.1 | - | 4 | 473 | 6401143 | ||
BC057592.1 | - | 4 | 578 | 6417362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
